Gene Transcripts

Gene Symbol
ASTL
Entrez Gene ID
431705
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Gene Aliases
SAS1B
Chromosome Location
Chr. 2: 96122817 - 96143059 on Build GRCh38
UniGene ID
Hs.447993
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011511205.2 XP_011509507.1 1 863
XM_011511207.2 XP_011509509.1 1 864
XM_011511208.2 XP_011509510.1 1 864

View Full Product Details