Gene Transcripts

Gene Symbol
OR4F3
Entrez Gene ID
26683
Gene Name
olfactory receptor family 4 subfamily F member 3
Gene Aliases
Chromosome Location
Chr. 5: 181342142 - 181370030 on Build GRCh38
UniGene ID
Hs.632360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005224.1 NP_001005224.1 1 233
XM_017009338.1 XP_016864827.1 2 4450
GenBank mRNA BC137547.1 - 1 252
BC137568.1 - 1 252 9021568

Gene Symbol
OR4F16
Entrez Gene ID
81399
Gene Name
olfactory receptor family 4 subfamily F member 16
Gene Aliases
OR1-1, OR7-21
Chromosome Location
Chr. 1: 683910 - 720118 on Build GRCh38
UniGene ID
Hs.632360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005277.1 NP_001005277.1 1 233
XM_017002408.1 XP_016857897.1 3 2965
XM_017002409.1 XP_016857898.1 4 2883
XM_017002410.1 XP_016857899.1 2 2747
XM_017002411.1 XP_016857900.1 2 2736

Gene Symbol
OR4F21
Entrez Gene ID
441308
Gene Name
olfactory receptor family 4 subfamily F member 21
Gene Aliases
OR4F21P
Chromosome Location
Chr. 8: 166086 - 167024 on Build GRCh38
UniGene ID
Hs.690459
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005504.1 NP_001005504.1 1 233

Gene Symbol
OR4F29
Entrez Gene ID
729759
Gene Name
olfactory receptor family 4 subfamily F member 29
Gene Aliases
OR7-21
Chromosome Location
Chr. 1: 450740 - 451678 on Build GRCh38
UniGene ID
Hs.632360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005221.2 NP_001005221.2 1 233

View Full Product Details