Gene Transcripts
- Gene Symbol
- FAM66A
- Entrez Gene ID
- 100133172
- Gene Name
- family with sequence similarity 66 member A
- Gene Aliases
- Chromosome Location
- Chr. 8: 12362019 - 12411001 on Build GRCh38
- UniGene ID
- Hs.721640
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026789.1 |
|
1 |
207 |
|
GenBank mRNA |
AK311040.1 |
- |
1 |
208 |
|
DC372067.1 |
- |
1 |
207 |
|
- Gene Symbol
- FAM66D
- Entrez Gene ID
- 100132923
- Gene Name
- family with sequence similarity 66 member D
- Gene Aliases
- Chromosome Location
- Chr. 8: 12115782 - 12151189 on Build GRCh38
- UniGene ID
- Hs.546711
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027425.1 |
|
1 |
207 |
|
GenBank mRNA |
AK294595.1 |
- |
1 |
207 |
|
- Gene Symbol
- FAM66E
- Entrez Gene ID
- 100132103
- Gene Name
- family with sequence similarity 66 member E
- Gene Aliases
- Chromosome Location
- Chr. 8: 7955013 - 8008755 on Build GRCh38
- UniGene ID
- Hs.694680
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027424.1 |
|
1 |
209 |
|
View Full Product Details