Gene Transcripts

Gene Symbol
SPANXN1
Entrez Gene ID
494118
Gene Name
SPANX family member N1
Gene Aliases
CT11.6
Chromosome Location
Chr. X: 145247587 - 145256208 on Build GRCh38
UniGene ID
Hs.551270
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009614.2 NP_001009614.1 2 252
GenBank mRNA DQ336118.1 - 2 252
DQ336119.1 - 2 252
DQ336120.1 - 2 252
DQ336121.1 - 2 252

Gene Symbol
SPANXN2
Entrez Gene ID
494119
Gene Name
SPANX family member N2
Gene Aliases
CT11.7, SPANX-N2
Chromosome Location
Chr. X: 143712035 - 143720668 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009615.2 NP_001009615.1 2 255
GenBank mRNA DQ336111.1 - 2 255
DQ336112.1 - 2 255
DQ336113.1 - 2 255
DQ336114.1 - 2 255
DQ336115.1 - 2 255

View Full Product Details