Gene Transcripts

Gene Symbol
C8orf58
Entrez Gene ID
541565
Gene Name
chromosome 8 open reading frame 58
Gene Aliases
Chromosome Location
Chr. 8: 22599601 - 22604150 on Build GRCh38
UniGene ID
Hs.743508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013842.2 NP_001013864.1 7 1170
NM_001198827.1 NP_001185756.1 6 1063
NM_173686.2 NP_775957.2 7 1146
GenBank mRNA AK092034.1 - 6 954
AK295405.1 - 7 1146
BC012750.2 - 6 961 3618901

Gene Symbol
LOC107986876
Entrez Gene ID
107986876
Gene Name
proline-rich proteoglycan 2-like
Gene Aliases
Chromosome Location
Chr. 8: 22602708 - 22605328 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017014122.1 XP_016869611.1 1 2067

View Full Product Details