Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198449.2 | NP_940851.1 | 2 | 383 | |
XM_011543146.2 | XP_011541448.1 | 3 | 165 | ||
GenBank mRNA | AK300860.1 | - | 2 | 348 | |
AK304226.1 | - | 2 | 254 | ||
BC059398.1 | - | 2 | 350 | 30332712 | |
DC309453.1 | - | 2 | 383 |