Gene Transcripts
- Gene Symbol
- SSBP3-AS1
- Entrez Gene ID
- 619518
- Gene Name
- SSBP3 antisense RNA 1
- Gene Aliases
- C1orf191, MST128, MSTP128
- Chromosome Location
- Chr. 1: 54236443 - 54239063 on Build GRCh38
- UniGene ID
- Hs.591438
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103541.1 |
|
1 |
1855 |
|
GenBank mRNA |
AF176918.1 |
- |
1 |
261 |
|
View Full Product Details