Gene Transcripts
- Gene Symbol
- TRAPPC3L
- Entrez Gene ID
- 100128327
- Gene Name
- trafficking protein particle complex 3 like
- Gene Aliases
- BET3L, bA259P20.2
- Chromosome Location
- Chr. 6: 116496483 - 116545610 on Build GRCh38
- UniGene ID
- Hs.134795
- Species
- Homo sapiens
- Gene Symbol
- FAM26D
- Entrez Gene ID
- 221301
- Gene Name
- family with sequence similarity 26 member D
- Gene Aliases
- C6orf78
- Chromosome Location
- Chr. 6: 116529013 - 116561123 on Build GRCh38
- UniGene ID
- Hs.350750
- Species
- Homo sapiens
View Full Product Details