Gene Transcripts
- Gene Symbol
- RPPH1
- Entrez Gene ID
- 85495
- Gene Name
- ribonuclease P RNA component H1
- Gene Aliases
- H1RNA, RPPH1-1
- Chromosome Location
- Chr. 14: 20343071 - 20343411 on Build GRCh38
- UniGene ID
- Hs.613102
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CA413366.1 |
- |
1 |
450 |
|
- Gene Symbol
- PARP2
- Entrez Gene ID
- 10038
- Gene Name
- poly(ADP-ribose) polymerase 2
- Gene Aliases
- ADPRT2, ADPRTL2, ADPRTL3, ARTD2, PARP-2, pADPRT-2
- Chromosome Location
- Chr. 14: 20343041 - 20357904 on Build GRCh38
- UniGene ID
- Hs.409412
- Species
- Homo sapiens
View Full Product Details