Gene Transcripts
- Gene Symbol
- ALYREF
- Entrez Gene ID
- 10189
- Gene Name
- Aly/REF export factor
- Gene Aliases
- ALY, ALY/REF, BEF, REF, THOC4
- Chromosome Location
- Chr. 17: 81887835 - 81891622 on Build GRCh38
- UniGene ID
- Hs.534385
- Species
- Homo sapiens
- Gene Symbol
- ANAPC11
- Entrez Gene ID
- 51529
- Gene Name
- anaphase promoting complex subunit 11
- Gene Aliases
- APC11, Apc11p, HSPC214
- Chromosome Location
- Chr. 17: 81890790 - 81900533 on Build GRCh38
- UniGene ID
- Hs.534456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BF435042.1 |
- |
1 |
77 |
|
BF512771.1 |
- |
1 |
84 |
|
View Full Product Details