Gene Transcripts
- Gene Symbol
- DDX19B
- Entrez Gene ID
- 11269
- Gene Name
- DEAD-box helicase 19B
- Gene Aliases
- DBP5, DDX19, RNAh
- Chromosome Location
- Chr. 16: 70289767 - 70335213 on Build GRCh38
- UniGene ID
- Hs.221761
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001014451.2 |
NP_001014451.1
|
1, 2 |
202 |
|
NM_007242.5 |
NP_009173.1
|
1, 2 |
202 |
|
XM_011522832.2 |
XP_011521134.1
|
2, 3 |
258 |
|
XM_011522833.2 |
XP_011521135.1
|
2, 3 |
258 |
|
GenBank mRNA |
AJ237946.1 |
- |
1, 2 |
63 |
|
AK027378.1 |
- |
1, 2 |
167 |
|
AK223115.1 |
- |
1, 2 |
173 |
|
AL136639.1 |
- |
1, 2 |
171 |
|
BC003626.2 |
- |
1, 2 |
166 |
3030038 |
CR457215.1 |
- |
1, 2 |
63 |
|
CR533509.1 |
- |
1, 2 |
63 |
|
DC385044.1 |
- |
1, 2 |
205 |
|
View Full Product Details