Gene Transcripts

Gene Symbol
PIH1D2
Entrez Gene ID
120379
Gene Name
PIH1 domain containing 2
Gene Aliases
Chromosome Location
Chr. 11: 112052161 - 112074288 on Build GRCh38
UniGene ID
Hs.420662
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082619.1 NP_001076088.1 3 479
NM_138789.3 NP_620144.1 3 479
XM_011542593.2 XP_011540895.2 3 385
XM_017017201.1 XP_016872690.1 3 596
XM_017017202.1 XP_016872691.1 3 596
XM_017017203.1 XP_016872692.1 3 596
XM_017017204.1 XP_016872693.1 3 596
XM_017017205.1 XP_016872694.1 3 596
GenBank mRNA AK300496.1 - 3 342
BC019238.2 - 2 288 3935322
CB159963.1 - 3 479

View Full Product Details