Gene Transcripts

Gene Symbol
TTC32
Entrez Gene ID
130502
Gene Name
tetratricopeptide repeat domain 32
Gene Aliases
Chromosome Location
Chr. 2: 19896631 - 19901986 on Build GRCh38
UniGene ID
Hs.591547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008237.2 NP_001008238.1 1 221
GenBank mRNA BC057850.1 - 1 221
BC070381.1 - 1 92

View Full Product Details