Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033505.1 | NP_001028677.1 | 2 | 267 | |
NM_001033506.1 | NP_001028678.1 | 2 | 267 | ||
NM_001326.2 | NP_001317.1 | 2 | 267 | ||
GenBank mRNA | AK290836.1 | - | 2 | 267 | |
BC009792.1 | - | 2 | 249 | ||
BC059948.1 | - | 2 | 121 | ||
BC108319.1 | - | 2 | 241 | 6197136 | |
BG435786.1 | - | 2 | 268 | ||
U15782.1 | - | 2 | 232 |