Gene Transcripts
- Gene Symbol
- CIART
- Entrez Gene ID
- 148523
- Gene Name
- circadian associated repressor of transcription
- Gene Aliases
- C1orf51, CHRONO, GM129
- Chromosome Location
- Chr. 1: 150282533 - 150287093 on Build GRCh38
- UniGene ID
- Hs.54680
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300838.1 |
NP_001287767.1
|
2, 3 |
502 |
|
NM_001300839.1 |
NP_001287768.1
|
2, 3 |
248 |
|
NM_001300840.1 |
NP_001287769.1
|
2, 3 |
196 |
|
NM_001300841.1 |
NP_001287770.1
|
2, 3 |
264 |
|
NM_144697.3 |
NP_653298.1
|
1, 2 |
1091 |
|
GenBank mRNA |
AK056960.1 |
- |
1, 2 |
372 |
|
AK315397.1 |
- |
1, 2 |
776 |
|
BC017397.1 |
- |
2, 3 |
220 |
|
BC027999.1 |
- |
2, 3 |
492 |
5194413 |
BC047238.1 |
- |
1, 2 |
805 |
5273646 |
BE784098.1 |
- |
2, 3 |
194 |
|
CN359562.1 |
- |
2, 3 |
162 |
|
View Full Product Details