Gene Transcripts

Gene Symbol
DPH2
Entrez Gene ID
1802
Gene Name
DPH2 homolog
Gene Aliases
DPH2L2
Chromosome Location
Chr. 1: 43969979 - 43973371 on Build GRCh38
UniGene ID
Hs.632398
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039589.1 NP_001034678.1 4, 5 854
NM_001319165.1 NP_001306094.1 5, 6 1528
NM_001319166.1 NP_001306095.1 5, 6 1265
NM_001319167.1 NP_001306096.1 5, 6 1595
NM_001319168.1 NP_001306097.1 5, 6 1486
NM_001319169.1 NP_001306098.1 4, 5 1314
NM_001319170.1 NP_001306099.1 5, 6 1598
NM_001319171.1 NP_001306100.1 5, 6 1569
NM_001384.4 NP_001375.2 5, 6 1538
GenBank mRNA AK091303.1 - 5, 6 1457
AK297933.1 - 4, 5 1287
AK315506.1 - 5, 6 1522
BC001389.2 - 5, 6 1497 3051455
BC003181.1 - 5, 6 1515 2958202
BC016956.1 - 5, 6 1497 3900854
BT007431.1 - 5, 6 1343
BX362196.2 - 1, 2 926

View Full Product Details