Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000214.2 | NP_000205.1 | 23 | 3392 | |
XM_017027707.1 | XP_016883196.1 | 22 | 3497 | ||
GenBank mRNA | AF003837.1 | - | 23 | 3335 | |
AF028593.1 | - | 23 | 3139 | ||
AK302554.1 | - | 21 | 2742 | ||
AK312722.1 | - | 23 | 2961 | ||
BC098393.1 | - | 23 | 3295 | 30528888 | |
BC126205.1 | - | 24 | 2900 | 8991921 | |
BC126207.1 | - | 23 | 3128 | 8991923 | |
U61276.1 | - | 23 | 3246 | ||
U73936.1 | - | 23 | 3289 | ||
U77720.1 | - | 23 | 2838 | ||
U77914.1 | - | 6 | 650 |