Gene Transcripts

Gene Symbol
ASF1A
Entrez Gene ID
25842
Gene Name
anti-silencing function 1A histone chaperone
Gene Aliases
CGI-98, CIA, HSPC146
Chromosome Location
Chr. 6: 118894077 - 118909171 on Build GRCh38
UniGene ID
Hs.292316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014034.2 NP_054753.1 1 364
GenBank mRNA AB028628.1 - 1 190
AF151856.1 - 3 641
AF161495.1 - 3 641
AF279306.1 - 1 176
AK025738.1 - 1 221
AL050261.1 - 1 219
BC010878.1 - 1 203 3546858
CR457347.1 - 1 27
DA902777.1 - 1 364

View Full Product Details