Gene Transcripts
- Gene Symbol
- EMC10
- Entrez Gene ID
- 284361
- Gene Name
- ER membrane protein complex subunit 10
- Gene Aliases
- C19orf63, HSM1, HSS1
- Chromosome Location
- Chr. 19: 50476477 - 50505905 on Build GRCh38
- UniGene ID
- Hs.448941
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_175063.5 |
NP_778233.4
|
3, 4 |
369 |
|
NM_206538.3 |
NP_996261.1
|
3, 4 |
369 |
|
XM_006723163.3 |
XP_006723226.1
|
3, 4 |
363 |
|
XM_011526818.2 |
XP_011525120.1
|
3, 4 |
363 |
|
GenBank mRNA |
AY194293.2 |
- |
7, 8 |
1327 |
|
AY358710.1 |
- |
3, 4 |
329 |
|
AY761095.1 |
- |
3, 4 |
347 |
|
AY761097.1 |
- |
3, 4 |
347 |
|
BC032948.2 |
- |
3, 4 |
310 |
4794992 |
BC035001.2 |
- |
3, 4 |
308 |
4796476 |
BC062607.1 |
- |
3, 4 |
511 |
|
BX366174.2 |
- |
3, 4 |
367 |
|
HY107516.1 |
- |
3, 4 |
370 |
|
View Full Product Details