Gene Transcripts

Gene Symbol
YIPF7
Entrez Gene ID
285525
Gene Name
Yip1 domain family member 7
Gene Aliases
FinGER9
Chromosome Location
Chr. 4: 44619578 - 44656868 on Build GRCh38
UniGene ID
Hs.596000
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182592.2 NP_872398.2 2 160
XM_011513679.2 XP_011511981.1 2 5387
GenBank mRNA BC103995.1 - 2 160
BC103996.1 - 2 160

View Full Product Details