Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001137673.1 | NP_001131145.1 | 3 | 369 | |
NM_004516.3 | NP_004507.2 | 3 | 369 | ||
NM_012218.3 | NP_036350.2 | 3 | 369 | ||
NM_017620.2 | NP_060090.2 | 3 | 369 | ||
NM_153464.2 | NP_703194.1 | 3 | 369 | ||
XM_006722742.2 | XP_006722805.1 | 3 | 284 | ||
XM_011527984.2 | XP_011526286.2 | 3 | 402 | ||
XM_011527985.2 | XP_011526287.1 | 3 | 284 | ||
XM_017026763.1 | XP_016882252.1 | 4 | 493 | ||
XM_017026764.1 | XP_016882253.1 | 1 | 49 | ||
XM_017026765.1 | XP_016882254.1 | 3 | 284 | ||
GenBank mRNA | AB209062.1 | - | 3 | 326 | |
AF141870.1 | - | 3 | 163 | ||
AF147209.1 | - | 1 | 64 | ||
AF167569.1 | - | 1 | 52 | ||
AF167570.1 | - | 1 | 52 | ||
AJ271745.1 | - | 2 | 141 | ||
AJ271746.1 | - | 2 | 121 | ||
AK291205.1 | - | 3 | 268 | ||
AK291617.1 | - | 3 | 254 | ||
AK294084.1 | - | 3 | 260 | ||
BC001770.1 | - | 3 | 254 | ||
BC048314.1 | - | 3 | 299 | ||
BC064836.1 | - | 3 | 302 | 6503234 | |
DC320902.1 | - | 3 | 372 | ||
HQ322263.1 | - | 1 | 52 | ||
HQ322264.1 | - | 1 | 52 | ||
HQ322265.1 | - | 1 | 52 | ||
U10324.1 | - | 3 | 316 | ||
X98264.1 | - | 3 | 256 | ||
X98265.1 | - | 2 | 261 |