Gene Transcripts
- Gene Symbol
- WASH1
- Entrez Gene ID
- 100287171
- Gene Name
- WAS protein family homolog 1
- Gene Aliases
- FAM39E, WASH
- Chromosome Location
- Chr. 9: 14475 - 30487 on Build GRCh38
- UniGene ID
- Hs.446466
- Species
- Homo sapiens
- Gene Symbol
- WASH3P
- Entrez Gene ID
- 374666
- Gene Name
- WAS protein family homolog 3 pseudogene
- Gene Aliases
- FAM39DP
- Chromosome Location
- Chr. 15: 101960813 - 101976605 on Build GRCh38
- UniGene ID
- Hs.459573
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK300161.1 |
- |
3, 4 |
395 |
|
- Gene Symbol
- WASH7P
- Entrez Gene ID
- 653635
- Gene Name
- WAS protein family homolog 7 pseudogene
- Gene Aliases
- FAM39F, WASH5P
- Chromosome Location
- Chr. 1: 14362 - 29370 on Build GRCh38
- UniGene ID
- Hs.446466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK294377.1 |
- |
1, 2 |
90 |
|
AK296182.1 |
- |
3, 4 |
330 |
|
AK316487.1 |
- |
1, 2 |
383 |
|
View Full Product Details