Gene Transcripts

Gene Symbol
ALDH7A1
Entrez Gene ID
501
Gene Name
aldehyde dehydrogenase 7 family member A1
Gene Aliases
ATQ1, EPD, PDE
Chromosome Location
Chr. 5: 126541841 - 126595390 on Build GRCh38
UniGene ID
Hs.483239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001182.4 NP_001173.2 13, 14 1398
NM_001201377.1 NP_001188306.1 13, 14 1398
XM_011543417.2 XP_011541719.1 13, 14 1389
XM_017009493.1 XP_016864982.1 13, 14 1174
GenBank mRNA AK092507.1 - 4, 5 1410
AK295526.1 - 8, 9 633
AK312459.1 - 13, 14 1227
BC002515.2 - 13, 14 1215 3139235
BC071712.1 - 13, 14 1192 5518212
BC073174.1 - 13, 14 1184 6501646
S74728.1 - 13, 14 1176

View Full Product Details