Gene Transcripts

Gene Symbol
TAS2R3
Entrez Gene ID
50831
Gene Name
taste 2 receptor member 3
Gene Aliases
T2R3
Chromosome Location
Chr. 7: 141764097 - 141765197 on Build GRCh38
UniGene ID
Hs.676011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016943.2 NP_058639.1 1 370
GenBank mRNA BC069337.1 - 1 308
BC095523.1 - 1 370 6971936

View Full Product Details