Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016298.3 | NP_057382.2 | 3 | 520 | |
GenBank mRNA | AB033021.1 | - | 3 | 724 | |
AF204674.1 | - | 3 | 426 | ||
AK314400.1 | - | 3 | 442 | ||
BC109275.1 | - | 2 | 331 | 40028315 | |
BC109276.1 | - | 2 | 331 | 40028318 | |
DB081227.1 | - | 3 | 520 |