Gene Transcripts

Gene Symbol
ACER3
Entrez Gene ID
55331
Gene Name
alkaline ceramidase 3
Gene Aliases
APHC, PHCA
Chromosome Location
Chr. 11: 76860873 - 77026797 on Build GRCh38
UniGene ID
Hs.23862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300954.1 NP_001287883.1 2 271
NM_001300955.1 NP_001287884.1 2 271
NM_018367.6 NP_060837.3 2 271
XM_011545151.2 XP_011543453.1 2 237
XM_011545152.2 XP_011543454.1 2 207
XM_011545153.2 XP_011543455.1 2 237
XM_017017987.1 XP_016873476.1 2 256
XM_017017988.1 XP_016873477.1 2 237
GenBank mRNA AF214454.1 - 2 225
AF327353.1 - 2 236
AK002100.1 - 2 209
AK293800.1 - 3 242
AK295142.1 - 2 188
AK295327.1 - 3 355
AK308856.1 - 3 257
AK315000.1 - 2 237
AK316120.1 - 2 209
BC063034.1 - 3 231
BC073853.1 - 2 188 3867010
DB461413.1 - 2 262

View Full Product Details