Gene Transcripts
- Gene Symbol
- CEACAM19
- Entrez Gene ID
- 56971
- Gene Name
- carcinoembryonic antigen related cell adhesion molecule 19
- Gene Aliases
- CEACM19, CEAL1
- Chromosome Location
- Chr. 19: 44671452 - 44684355 on Build GRCh38
- UniGene ID
- Hs.416925
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127893.1 |
NP_001121365.1
|
5, 6 |
1187 |
|
NM_020219.3 |
NP_064604.2
|
5, 6 |
1187 |
|
GenBank mRNA |
AK027157.1 |
- |
5, 6 |
904 |
|
AK128234.1 |
- |
2, 3 |
1398 |
|
AK291588.1 |
- |
5, 6 |
888 |
|
AY358394.1 |
- |
5, 6 |
917 |
|
BC083499.1 |
- |
5, 6 |
1187 |
30390224 |
KJ851728.1 |
- |
5, 6 |
707 |
|
KJ851729.1 |
- |
5, 6 |
707 |
|
KJ851730.1 |
- |
5, 6 |
707 |
|
KJ851731.1 |
- |
5, 6 |
707 |
|
KJ851732.1 |
- |
4, 5 |
623 |
|
KJ851733.1 |
- |
4, 5 |
623 |
|
KJ851736.1 |
- |
4, 5 |
623 |
|
KJ851737.1 |
- |
4, 5 |
623 |
|
KJ851738.1 |
- |
4, 5 |
623 |
|
KJ851739.1 |
- |
4, 5 |
623 |
|
KJ851742.1 |
- |
4, 5 |
556 |
|
KJ851743.1 |
- |
4, 5 |
556 |
|
KJ851746.1 |
- |
4, 5 |
556 |
|
KJ851747.1 |
- |
4, 5 |
556 |
|
KJ851748.1 |
- |
4, 5 |
556 |
|
KJ851749.1 |
- |
4, 5 |
556 |
|
KJ851750.1 |
- |
3, 4 |
472 |
|
KJ851751.1 |
- |
3, 4 |
472 |
|
KJ851754.1 |
- |
3, 4 |
472 |
|
KJ851755.1 |
- |
3, 4 |
472 |
|
KJ851756.1 |
- |
3, 4 |
472 |
|
KJ851757.1 |
- |
3, 4 |
472 |
|
KJ851760.1 |
- |
6, 7 |
841 |
|
KJ851761.1 |
- |
6, 7 |
841 |
|
KJ851764.1 |
- |
6, 7 |
841 |
|
KJ851765.1 |
- |
6, 7 |
841 |
|
KJ851766.1 |
- |
6, 7 |
841 |
|
KJ851767.1 |
- |
6, 7 |
841 |
|
KJ851768.1 |
- |
5, 6 |
757 |
|
KJ851769.1 |
- |
5, 6 |
757 |
|
KJ851772.1 |
- |
5, 6 |
757 |
|
KJ851773.1 |
- |
5, 6 |
757 |
|
KJ851774.1 |
- |
5, 6 |
757 |
|
KJ851775.1 |
- |
5, 6 |
757 |
|
KJ851778.1 |
- |
5, 6 |
690 |
|
KJ851779.1 |
- |
5, 6 |
690 |
|
KJ851782.1 |
- |
5, 6 |
690 |
|
KJ851783.1 |
- |
5, 6 |
690 |
|
KJ851784.1 |
- |
5, 6 |
690 |
|
KJ851785.1 |
- |
5, 6 |
690 |
|
KJ851786.1 |
- |
4, 5 |
606 |
|
KJ851787.1 |
- |
4, 5 |
606 |
|
KJ851790.1 |
- |
4, 5 |
606 |
|
KJ851791.1 |
- |
4, 5 |
606 |
|
KJ851792.1 |
- |
4, 5 |
606 |
|
KJ851793.1 |
- |
4, 5 |
606 |
|
View Full Product Details