Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000986.3 | NP_000977.1 | 4 | 278 | |
GenBank mRNA | AK311992.1 | - | 4 | 282 | |
BC000690.2 | - | 4 | 257 | 3349215 | |
BC070193.1 | - | 4 | 255 | 6733162 | |
BU754101.1 | - | 4 | 286 | ||
CD513036.1 | - | 3 | 264 | ||
CR456729.1 | - | 3 | 235 | ||
EU668341.1 | - | 4 | 257 | ||
M94314.1 | - | 4 | 274 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_011518475.2 | XP_011516777.1 | 12 | 2837 | |
GenBank mRNA | BX538309.1 | - | 1 | 2352 |