Gene Transcripts

Gene Symbol
GRXCR2
Entrez Gene ID
643226
Gene Name
glutaredoxin and cysteine rich domain containing 2
Gene Aliases
DFNB101
Chromosome Location
Chr. 5: 145858538 - 145931673 on Build GRCh38
UniGene ID
Hs.570895
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080516.1 NP_001073985.1 1 81
GenBank mRNA DN831963.1 - 1 19

View Full Product Details