Gene Transcripts

Gene Symbol
SHH
Entrez Gene ID
6469
Gene Name
sonic hedgehog
Gene Aliases
HHG1, HLP3, HPE3, MCOPCB5, SMMCI, TPT, TPTPS
Chromosome Location
Chr. 7: 155799984 - 155812273 on Build GRCh38
UniGene ID
Hs.164537
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000193.3 NP_000184.1 1 406
GenBank mRNA BX452210.1 - 1 311
L38518.1 - 1 406

View Full Product Details