Gene Transcripts
- Gene Symbol
- EPS8L2
- Entrez Gene ID
- 64787
- Gene Name
- EPS8 like 2
- Gene Aliases
- EPS8R2
- Chromosome Location
- Chr. 11: 705217 - 727727 on Build GRCh38
- UniGene ID
- Hs.55016
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022772.3 |
NP_073609.2
|
12, 13 |
1300 |
|
XM_017018130.1 |
XP_016873619.1
|
13, 14 |
1517 |
|
XM_017018131.1 |
XP_016873620.1
|
12, 13 |
1388 |
|
XM_017018132.1 |
XP_016873621.1
|
13, 14 |
1358 |
|
GenBank mRNA |
AK025588.1 |
- |
12, 13 |
1218 |
|
AK025824.1 |
- |
12, 13 |
1218 |
|
AK094539.1 |
- |
4, 5 |
396 |
|
AK122903.1 |
- |
9, 10 |
1496 |
|
AK222903.1 |
- |
12, 13 |
1213 |
|
AK225311.1 |
- |
12, 13 |
1218 |
|
AY074929.1 |
- |
12, 13 |
1325 |
|
BC080636.1 |
- |
12, 13 |
1304 |
6572993 |
BC093878.1 |
- |
11, 12 |
1093 |
7939723 |
BC101481.1 |
- |
11, 12 |
1093 |
8068987 |
BC143242.1 |
- |
11, 12 |
1141 |
9051743 |
View Full Product Details