Gene Transcripts

Gene Symbol
SHISA9
Entrez Gene ID
729993
Gene Name
shisa family member 9
Gene Aliases
CKAMP44
Chromosome Location
Chr. 16: 12901619 - 13563427 on Build GRCh38
UniGene ID
Hs.130661
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145204.2 NP_001138676.2 4, 5 1350
XM_005255539.3 XP_005255596.2 3, 4 1303
GenBank mRNA AK299610.1 - 5, 6 780

View Full Product Details