Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032119.3 | NP_115495.3 | 74 | 15165 | |
NR_003149.1 | 74 | 15082 | |||
XM_017009963.1 | XP_016865452.1 | 74 | 15305 | ||
XM_017009964.1 | XP_016865453.1 | 74 | 15302 | ||
XM_017009965.1 | XP_016865454.1 | 74 | 15187 | ||
XM_017009966.1 | XP_016865455.1 | 73 | 15224 | ||
XM_017009967.1 | XP_016865456.1 | 73 | 15094 | ||
XM_017009968.1 | XP_016865457.1 | 73 | 15125 | ||
XM_017009969.1 | XP_016865458.1 | 74 | 15305 | ||
XM_017009971.1 | XP_016865460.1 | 73 | 15130 | ||
XM_017009972.1 | XP_016865461.1 | 46 | 8587 | ||
XM_017009973.1 | XP_016865462.1 | 51 | 9798 | ||
GenBank mRNA | AF055084.1 | - | 10 | 2336 | |
AF435925.1 | - | 74 | 15168 | ||
AK024416.1 | - | 4 | 541 |