Gene Transcripts
- Gene Symbol
- TXNL1
- Entrez Gene ID
- 9352
- Gene Name
- thioredoxin like 1
- Gene Aliases
- HEL-S-114, TRP32, TXL-1, TXNL, Txl
- Chromosome Location
- Chr. 18: 56602822 - 56651276 on Build GRCh38
- UniGene ID
- Hs.114412
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004786.2 |
NP_004777.1
|
2, 3 |
452 |
|
NR_024546.1 |
|
3, 4 |
557 |
|
XM_006722580.3 |
XP_006722643.1
|
3, 4 |
1057 |
|
XM_011526263.2 |
XP_011524565.1
|
4, 5 |
1149 |
|
XM_011526264.2 |
XP_011524566.1
|
3, 4 |
296 |
|
XM_017026093.1 |
XP_016881582.1
|
4, 5 |
1149 |
|
XM_017026094.1 |
XP_016881583.1
|
3, 4 |
296 |
|
GenBank mRNA |
AB209263.1 |
- |
2, 3 |
190 |
|
AF003938.1 |
- |
2, 3 |
408 |
|
AF051896.1 |
- |
2, 3 |
333 |
|
AF052659.1 |
- |
2, 3 |
352 |
|
AK094926.1 |
- |
4, 5 |
1166 |
|
AK313653.1 |
- |
2, 3 |
363 |
|
BC001156.1 |
- |
2, 3 |
396 |
3534029 |
DA248363.1 |
- |
3, 4 |
296 |
|
FJ224353.1 |
- |
2, 3 |
408 |
|
View Full Product Details