The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260509.1 | NP_001247438.1 | 1 - 2 | 898 | 115 |
NM_002239.3 | NP_002230.1 | 1 - 2 | 898 | 115 | |
GenBank mRNA | BC022495.1 | - | 1 - 2 | 733 | 115 |
GU068048.1 | - | 1 - 2 | 727 | 115 | |
U50964.1 | - | 1 - 2 | 899 | 115 |