The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002864.2 | NP_002855.2 | 1 - 2 | 115 | 109 |
XM_011520804.2 | XP_011519106.1 | 1 - 2 | 114 | 109 | |
XM_011520805.2 | XP_011519107.1 | 1 - 2 | 114 | 109 | |
XM_011520806.1 | XP_011519108.1 | 1 - 2 | 114 | 109 | |
XM_011520807.2 | XP_011519109.1 | 1 - 2 | 114 | 109 | |
XM_017019763.1 | XP_016875252.1 | 1 - 2 | 114 | 109 | |
XM_017019764.1 | XP_016875253.1 | 1 - 2 | 114 | 109 | |
XM_017019765.1 | XP_016875254.1 | 1 - 2 | 114 | 109 | |
XM_017019766.1 | XP_016875255.1 | 1 - 2 | 110 | 109 | |
XM_017019767.1 | XP_016875256.1 | 1 - 2 | 114 | 109 | |
GenBank mRNA | AK313639.1 | - | 1 - 2 | 114 | 109 |
BC111756.1 | - | 1 - 2 | 110 | 109 | |
X54380.1 | - | 1 - 2 | 115 | 109 |