The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003245.3 | NP_003236.3 | 5 - 6 | 768 | 76 |
GenBank mRNA | AK290324.1 | - | 5 - 6 | 732 | 76 |
AK291351.1 | - | 5 - 6 | 732 | 76 | |
AK298637.1 | - | 5 - 6 | 734 | 76 | |
AK308249.1 | - | 5 - 6 | 734 | 76 | |
AK315236.1 | - | 5 - 6 | 732 | 76 | |
AL691671.1 | - | 1 - 2 | 51 | 76 | |
BC109075.1 | - | 5 - 6 | 694 | 76 | |
BC109076.1 | - | 5 - 6 | 694 | 76 | |
L10386.1 | - | 5 - 6 | 712 | 76 |