Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACVRL1
Entrez Gene ID
94
Gene Name
activin A receptor like type 1
Gene Aliases
ACVRLK1, ALK-1, ALK1, HHT, HHT2, ORW2, SKR3, TSR-I
Chromosome Location
Chr.12: 51906913 - 51923361 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000020.2 NP_000011.2 1 - 2 283 137
XM_005269235.2 XP_005269292.1 2 - 3 285 137
GenBank mRNA DN999570.1 - 1 - 2 291 137
HM161905.1 - 2 - 3 290 137
HM161906.1 - 2 - 3 250 137
Z22533.1 - 1 - 2 282 137

View Full Product Details