The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000346.3 | NP_000337.1 | 2 - 3 | 1060 | 102 |
GenBank mRNA | AK295455.1 | - | 2 - 3 | 1057 | 102 |
BC007951.2 | - | 2 - 3 | 917 | 102 | |
BC056420.1 | - | 2 - 3 | 1070 | 102 | |
BT006875.1 | - | 2 - 3 | 688 | 102 | |
Z46629.1 | - | 2 - 3 | 1047 | 102 |