The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000428.2 | NP_000419.1 | 18 - 19 | 3296 | 78 |
XM_011536765.1 | XP_011535067.1 | 16 - 17 | 2823 | 78 | |
GenBank mRNA | AB209865.1 | - | 18 - 19 | 2546 | 78 |
BC078659.1 | - | 19 - 20 | 3202 | 78 | |
S82451.1 | - | 18 - 19 | 3296 | 78 | |
Z37976.1 | - | 18 - 19 | 3297 | 78 |