Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- UGT1A4
- Entrez Gene ID
-
54657
- Gene Name
- UDP glucuronosyltransferase family 1 member A4
- Gene Aliases
-
HUG-BR2, UDPGT, UDPGT 1-4, UGT-1D, UGT1-04, UGT1.4, UGT1A4S, UGT1D
- Chromosome Location
-
Chr.2: 233718792 - 233773299 on Build GRCh38
- Gene Symbol
- UGT1A1
- Entrez Gene ID
-
54658
- Gene Name
- UDP glucuronosyltransferase family 1 member A1
- Gene Aliases
-
BILIQTL1, GNT1, HUG-BR1, UDPGT, UDPGT 1-1, UGT1, UGT1A
- Chromosome Location
-
Chr.2: 233760273 - 233773299 on Build GRCh38
- Gene Symbol
- UGT1A10
- Entrez Gene ID
-
54575
- Gene Name
- UDP glucuronosyltransferase family 1 member A10
- Gene Aliases
-
UDPGT, UGT-1J, UGT1-10, UGT1.10, UGT1J
- Chromosome Location
-
Chr.2: 233636477 - 233773305 on Build GRCh38
- Gene Symbol
- UGT1A3
- Entrez Gene ID
-
54659
- Gene Name
- UDP glucuronosyltransferase family 1 member A3
- Gene Aliases
-
UDPGT, UDPGT 1-3, UGT-1C, UGT1-03, UGT1.3, UGT1A3S, UGT1C
- Chromosome Location
-
Chr.2: 233729127 - 233773299 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_019093.2 |
NP_061966.1 |
4 - 5 |
1310 |
98 |
GenBank mRNA |
AY435138.1 |
- |
4 - 5 |
1310 |
98 |
- Gene Symbol
- UGT1A5
- Entrez Gene ID
-
54579
- Gene Name
- UDP glucuronosyltransferase family 1 member A5
- Gene Aliases
-
UDPGT, UDPGT 1-5, UGT1E
- Chromosome Location
-
Chr.2: 233712992 - 233773299 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_019078.1 |
NP_061951.1 |
4 - 5 |
1310 |
98 |
GenBank mRNA |
AY435140.1 |
- |
4 - 5 |
1310 |
98 |
- Gene Symbol
- UGT1A6
- Entrez Gene ID
-
54578
- Gene Name
- UDP glucuronosyltransferase family 1 member A6
- Gene Aliases
-
GNT1, HLUGP, HLUGP1, UDPGT, UDPGT 1-6, UGT1, UGT1A6S, UGT1F
- Chromosome Location
-
Chr.2: 233691675 - 233773305 on Build GRCh38
- Gene Symbol
- UGT1A7
- Entrez Gene ID
-
54577
- Gene Name
- UDP glucuronosyltransferase family 1 member A7
- Gene Aliases
-
UDPGT, UDPGT 1-7, UGT-1G, UGT1-07, UGT1.7, UGT1G
- Chromosome Location
-
Chr.2: 233681938 - 233773299 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_019077.2 |
NP_061950.2 |
4 - 5 |
1298 |
98 |
GenBank mRNA |
AY435142.1 |
- |
4 - 5 |
1298 |
98 |
- Gene Symbol
- UGT1A8
- Entrez Gene ID
-
54576
- Gene Name
- UDP glucuronosyltransferase family 1 member A8
- Gene Aliases
-
UDPGT, UDPGT 1-8, UGT-1H, UGT1-08, UGT1.8, UGT1A8S, UGT1H
- Chromosome Location
-
Chr.2: 233617645 - 233773299 on Build GRCh38
- Gene Symbol
- UGT1A9
- Entrez Gene ID
-
54600
- Gene Name
- UDP glucuronosyltransferase family 1 member A9
- Gene Aliases
-
HLUGP4, LUGP4, UDPGT, UDPGT 1-9, UGT-1I, UGT1-09, UGT1-9, UGT1.9, UGT1A9S, UGT1AI, UGT1I
- Chromosome Location
-
Chr.2: 233671898 - 233773305 on Build GRCh38
View Full Product Details