The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000620.4 | NP_000611.1 | 18 - 19 | 3538 | 61 |
NM_001204213.1 | NP_001191142.1 | 17 - 18 | 2130 | 61 | |
NM_001204214.1 | NP_001191143.1 | 17 - 18 | 2125 | 61 | |
NM_001204218.1 | NP_001191147.1 | 19 - 20 | 3640 | 61 | |
XM_011538398.2 | XP_011536700.1 | 20 - 21 | 3511 | 61 | |
XM_017019345.1 | XP_016874834.1 | 19 - 20 | 3647 | 61 | |
XM_017019346.1 | XP_016874835.1 | 19 - 20 | 3461 | 61 | |
XM_017019347.1 | XP_016874836.1 | 18 - 19 | 3448 | 61 | |
GenBank mRNA | AK294435.1 | - | 18 - 19 | 3342 | 61 |
AK307481.1 | - | 18 - 19 | 3538 | 61 | |
D16408.1 | - | 17 - 18 | 2827 | 61 | |
EU753241.1 | - | 18 - 19 | 3512 | 61 | |
L02881.1 | - | 17 - 18 | 3254 | 61 | |
U17327.1 | - | 18 - 19 | 3512 | 61 | |
U31466.1 | - | 17 - 18 | 2834 | 61 |