Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNLIP
Entrez Gene ID
5406
Gene Name
pancreatic lipase
Gene Aliases
PL, PNLIPD, PTL
Chromosome Location
Chr.10: 116545916 - 116567855 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA A24059.1 - 3 - 4 437 64
AK313941.1 - 5 - 6 489 64
BC014309.2 - 4 - 5 461 64
J05125.1 - 4 - 5 473 64
M93285.1 - 4 - 5 461 64

View Full Product Details