The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286555.1 | NP_001273484.1 | 1 - 2 | 505 | 109 |
NM_020185.4 | NP_064570.1 | 1 - 2 | 505 | 109 | |
NR_104473.1 | 1 - 2 | 505 | 109 | ||
XM_017011061.1 | XP_016866550.1 | 1 - 2 | 103 | 109 | |
GenBank mRNA | AF165519.1 | - | 1 - 2 | 46 | 109 |
AF424702.1 | - | 1 - 2 | 105 | 109 | |
AK296402.1 | - | 1 - 2 | 75 | 109 | |
AK310653.1 | - | 1 - 2 | 85 | 109 | |
AY249859.1 | - | 1 - 2 | 509 | 109 | |
BC022847.2 | - | 1 - 2 | 461 | 109 | |
DA950312.1 | - | 1 - 2 | 88 | 109 |