The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004357.4 | NP_004348.2 | 2 - 3 | 142 | 67 |
NM_139029.1 | NP_620598.1 | 2 - 3 | 138 | 67 | |
GenBank mRNA | AK130369.1 | - | 2 - 3 | 181 | 67 |
AK223186.1 | - | 2 - 3 | 85 | 67 | |
AK293073.1 | - | 2 - 3 | 109 | 67 | |
AL161965.1 | - | 2 - 3 | 104 | 67 | |
AU099249.1 | - | 2 - 3 | 142 | 67 | |
BC001374.2 | - | 1 - 2 | 81 | 67 | |
BC013302.2 | - | 2 - 3 | 114 | 67 | |
D29963.1 | - | 1 - 2 | 82 | 67 |