The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005076.3 | NP_005067.1 | 1 - 2 | 188 | 123 |
XM_017002199.1 | XP_016857688.1 | 1 - 2 | 240 | 123 | |
GenBank mRNA | CN289415.1 | - | 1 - 2 | 202 | 123 |
X67734.1 | - | 1 - 2 | 100 | 123 |