The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011705.1 | NP_001011705.1 | 4 - 5 | 581 | 86 |
NM_007015.2 | NP_008946.1 | 4 - 5 | 581 | 86 | |
XM_011534897.2 | XP_011533199.1 | 4 - 5 | 579 | 86 | |
XM_011534898.2 | XP_011533200.1 | 4 - 5 | 580 | 86 | |
GenBank mRNA | AB006000.1 | - | 4 - 5 | 470 | 86 |
BC025659.1 | - | 4 - 5 | 581 | 86 | |
CR541910.1 | - | 4 - 5 | 470 | 86 |