The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002987.2 | NP_002978.1 | 2 - 3 | 200 | 51 |
XM_011523256.2 | XP_011521558.1 | 4 - 5 | 571 | 51 | |
XM_017023530.1 | XP_016879019.1 | 4 - 5 | 574 | 51 | |
GenBank mRNA | BC069107.1 | - | 2 - 3 | 200 | 51 |
BC112066.1 | - | 2 - 3 | 166 | 51 | |
BC112068.1 | - | 2 - 3 | 169 | 51 | |
D43767.1 | - | 1 - 2 | 123 | 51 |