The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002994.4 | NP_002985.1 | 3 - 4 | 470 | 90 |
GenBank mRNA | BC008376.1 | - | 3 - 4 | 421 | 90 |
BU169770.1 | - | 3 - 4 | 441 | 90 | |
X78686.1 | - | 3 - 4 | 428 | 90 |