The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000131.4 | NP_000122.1 | 3 - 4 | 344 | 64 |
NM_019616.3 | NP_062562.1 | 2 - 3 | 278 | 64 | |
XM_006719963.3 | XP_006720026.2 | 2 - 3 | 293 | 64 | |
XM_011537474.2 | XP_011535776.2 | 2 - 3 | 293 | 64 | |
GenBank mRNA | AK310779.1 | - | 2 - 3 | 278 | 64 |
AK311565.1 | - | 2 - 3 | 193 | 64 | |
BC130468.1 | - | 2 - 3 | 268 | 64 | |
DB184045.1 | - | 2 - 3 | 278 | 64 | |
EU557239.1 | - | 3 - 4 | 294 | 64 | |
M13232.1 | - | 3 - 4 | 325 | 64 |